Assay Design



Gene Transcripts

Gene Symbol
Pigo
Entrez Gene ID
313341
Gene Name
phosphatidylinositol glycan anchor biosynthesis, class O
Gene Aliases
-
Chromosome Location
Chr.5: 58461055 - 58470693 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006238127.2 XP_006238189.1 6 - 7 1234 66
XM_233141.8 XP_233141.7 6 - 7 1777 66

View Full Product Details