Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191755.1 | NP_001178684.1 | 6 - 7 | 1111 | 158 |
XM_006237890.2 | XP_006237952.1 | 7 - 8 | 2916 | 158 | |
XM_006237891.1 | XP_006237953.1 | 5 - 6 | 2964 | 158 | |
XM_006237892.2 | XP_006237954.1 | 7 - 8 | 2916 | 158 | |
XM_008763543.1 | XP_008761765.1 | 7 - 8 | 2916 | 158 |