Assay Design



Gene Transcripts

Gene Symbol
Prim2
Entrez Gene ID
301323
Gene Name
primase, DNA, polypeptide 2
Gene Aliases
Pola3
Chromosome Location
Chr.9: 38535439 - 38745058 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006244686.2 XP_006244748.1 12 - 13 1384 83
XM_006244689.2 XP_006244751.1 12 - 13 1335 83
XM_006244690.2 XP_006244752.1 12 - 13 1401 83
XM_006244691.1 XP_006244753.1 12 - 13 1402 83
GenBank mRNA AJ011607.1 - 11 - 12 1239 83

View Full Product Details