Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024762.1 | NP_001019933.1 | 5 - 6 | 583 | 67 |
XM_006244686.2 | XP_006244748.1 | 3 - 4 | 412 | 67 | |
XM_006244689.2 | XP_006244751.1 | 3 - 4 | 363 | 67 | |
XM_006244690.2 | XP_006244752.1 | 3 - 4 | 429 | 67 | |
XM_006244691.1 | XP_006244753.1 | 3 - 4 | 430 | 67 | |
GenBank mRNA | AJ011607.1 | - | 2 - 3 | 267 | 67 |
BC097454.1 | - | 5 - 6 | 583 | 67 |