Assay Design



Gene Transcripts

Gene Symbol
Prim2
Entrez Gene ID
301323
Gene Name
primase, DNA, polypeptide 2
Gene Aliases
Pola3
Chromosome Location
Chr.9: 38535439 - 38745058 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024762.1 NP_001019933.1 7 - 8 779 89
XM_006244686.2 XP_006244748.1 5 - 6 608 89
XM_006244689.2 XP_006244751.1 5 - 6 559 89
XM_006244690.2 XP_006244752.1 5 - 6 625 89
XM_006244691.1 XP_006244753.1 5 - 6 626 89
GenBank mRNA AJ011607.1 - 4 - 5 463 89
BC097454.1 - 7 - 8 779 89

View Full Product Details