Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024762.1 | NP_001019933.1 | 9 - 10 | 1014 | 90 |
XM_006244686.2 | XP_006244748.1 | 7 - 8 | 843 | 90 | |
XM_006244689.2 | XP_006244751.1 | 7 - 8 | 794 | 90 | |
XM_006244690.2 | XP_006244752.1 | 7 - 8 | 860 | 90 | |
XM_006244691.1 | XP_006244753.1 | 7 - 8 | 861 | 90 | |
GenBank mRNA | AJ011607.1 | - | 6 - 7 | 698 | 90 |
BC097454.1 | - | 9 - 10 | 1014 | 90 |