Assay Design



Gene Transcripts

Gene Symbol
Bzw1
Entrez Gene ID
363232
Gene Name
basic leucine zipper and W2 domains 1
Gene Aliases
-
Chromosome Location
Chr.9: 65279675 - 65293471 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198789.2 NP_942084.1 4 - 5 437 116
XM_006245000.2 XP_006245062.2 4 - 5 436 116
GenBank mRNA BC061580.1 - 4 - 5 441 116

View Full Product Details