Assay Design



Gene Transcripts

Gene Symbol
Plekhb2
Entrez Gene ID
301337
Gene Name
pleckstrin homology domain containing, family B (evectins) member 2
Gene Aliases
-
Chromosome Location
Chr.9: 41348447 - 41381092 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106899.1 NP_001100369.1 2 - 3 145 114

View Full Product Details