Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106555.1 | NP_001100025.1 | 2 - 3 | 329 | 76 |
XM_006233581.2 | XP_006233643.1 | 2 - 3 | 714 | 76 | |
GenBank mRNA | FQ210613.1 | - | 2 - 3 | 289 | 76 |
FQ212796.1 | - | 2 - 3 | 576 | 76 | |
FQ218819.1 | - | 2 - 3 | 289 | 76 |