Assay Design



Gene Transcripts

Gene Symbol
Phpt1
Entrez Gene ID
296571
Gene Name
phosphohistidine phosphatase 1
Gene Aliases
-
Chromosome Location
Chr.3: 2797037 - 2798436 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106558.2 NP_001100028.2 2 - 3 301 116
GenBank mRNA BI276399.1 - 1 - 2 116
BI276995.1 - 1 - 2 116

View Full Product Details