Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004080.1 | NP_001004080.1 | 3 - 4 | 511 | 90 |
XM_006234039.2 | XP_006234101.1 | 3 - 4 | 409 | 90 | |
XM_006234040.1 | XP_006234102.1 | 4 - 5 | 529 | 90 | |
GenBank mRNA | BC079472.1 | - | 3 - 4 | 511 | 90 |