Assay Design



Gene Transcripts

Gene Symbol
Gsn
Entrez Gene ID
296654
Gene Name
gelsolin
Gene Aliases
-
Chromosome Location
Chr.3: 14456114 - 14508922 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004080.1 NP_001004080.1 6 - 7 908 75
XM_006234039.2 XP_006234101.1 6 - 7 806 75
XM_006234040.1 XP_006234102.1 7 - 8 926 75
GenBank mRNA BC079472.1 - 6 - 7 908 75
FQ223877.1 - 7 - 8 931 75

View Full Product Details