Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271240.1 | NP_001258169.1 | 5 - 6 | 932 | 78 |
XM_006233999.2 | XP_006234061.1 | 8 - 9 | 1289 | 78 | |
XM_006234000.2 | XP_006234062.1 | 8 - 9 | 1204 | 78 | |
GenBank mRNA | BC158696.1 | - | 8 - 9 | 1076 | 78 |