Assay Design



Gene Transcripts

Gene Symbol
Tmem8c
Entrez Gene ID
296601
Gene Name
transmembrane protein 8C
Gene Aliases
RGD1306074
Chromosome Location
Chr.3: 5608243 - 5617689 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134517.1 NP_001127989.1 1 - 2 326 86
XM_008761635.1 XP_008759857.1 1 - 2 328 86

View Full Product Details