Assay Design



Gene Transcripts

Gene Symbol
Ropn1
Entrez Gene ID
288053
Gene Name
rhophilin associated tail protein 1
Gene Aliases
-
Chromosome Location
Chr.11: 69326785 - 69355788 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025628.1 NP_001020799.1 2 - 3 270 89
XM_006248431.2 XP_006248493.1 2 - 3 213 89
GenBank mRNA BC099132.1 - 2 - 3 270 89

View Full Product Details