Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024344.1 | NP_001019515.1 | 3 - 4 | 347 | 91 |
GenBank mRNA | BC088199.1 | - | 3 - 4 | 347 | 91 |
FQ225852.1 | - | 2 - 3 | 262 | 91 | |
FQ233921.1 | - | 2 - 3 | 262 | 91 |