Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108663.1 | NP_001102133.1 | 7 - 8 | 1177 | 94 |
XM_006238084.1 | XP_006238146.1 | 7 - 8 | 1099 | 94 | |
XM_006238085.2 | XP_006238147.1 | 7 - 8 | 1256 | 94 | |
XM_006238086.1 | XP_006238148.1 | 6 - 7 | 992 | 94 | |
XM_006238088.2 | XP_006238150.1 | 7 - 8 | 1099 | 94 | |
XM_006238089.2 | XP_006238151.1 | 7 - 8 | 1099 | 94 | |
XM_006238090.2 | XP_006238152.1 | 7 - 8 | 1099 | 94 | |
XM_008763698.1 | XP_008761920.1 | 7 - 8 | 1099 | 94 |