Assay Design



Gene Transcripts

Gene Symbol
Map7
Entrez Gene ID
293016
Gene Name
microtubule-associated protein 7
Gene Aliases
Mtap7
Chromosome Location
Chr.1: 15620665 - 15748203 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106270.2 NP_001099740.2 11 - 12 1703 65
NM_001198638.1 NP_001185567.1 9 - 10 1236 65
GenBank mRNA CK480167.1 - 2 - 3 424 65

View Full Product Details