Assay Design



Gene Transcripts

Gene Symbol
Ltv1
Entrez Gene ID
361452
Gene Name
LTV1 ribosome biogenesis factor
Gene Aliases
RGD1311264
Chromosome Location
Chr.1: 7323043 - 7335766 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014157.1 NP_001014179.1 6 - 7 791 150
GenBank mRNA BC079397.1 - 6 - 7 791 150
FQ213117.1 - 1 - 2 227 150

View Full Product Details