Assay Design



Gene Transcripts

Gene Symbol
Ltv1
Entrez Gene ID
361452
Gene Name
LTV1 ribosome biogenesis factor
Gene Aliases
RGD1311264
Chromosome Location
Chr.1: 7323043 - 7335766 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014157.1 NP_001014179.1 9 - 10 1121 95
GenBank mRNA BC079397.1 - 9 - 10 1121 95
FQ213117.1 - 4 - 5 557 95

View Full Product Details