Assay Design



Gene Transcripts

Gene Symbol
Grm1
Entrez Gene ID
24414
Gene Name
glutamate receptor, metabotropic 1
Gene Aliases
Gprc1a
Chromosome Location
Chr.1: 4753144 - 5165859 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017011.1 NP_058707.1 8 - 9 3035 82
GenBank mRNA M61099.1 - 8 - 9 3035 82
X57569.1 - 8 - 9 3017 82
Y18812.1 - 1 - 2 162 82

View Full Product Details