Assay Design



Gene Transcripts

Gene Symbol
Aig1
Entrez Gene ID
292486
Gene Name
androgen-induced 1
Gene Aliases
RGD1562920
Chromosome Location
Chr.1: 7816841 - 8038808 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006227642.2 XP_006227704.1 5 - 6 907 128

View Full Product Details