Assay Design



Gene Transcripts

Gene Symbol
Cd99
Entrez Gene ID
652929
Gene Name
CD99 molecule
Gene Aliases
-
Chromosome Location
Chr.20: 56192426 - 56196961 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100804.1 NP_001094274.1 5 - 6 284 90
GenBank mRNA AY262357.1 - 5 - 6 273 90
BC168196.1 - - 284 90

View Full Product Details