Assay Design



Gene Transcripts

Gene Symbol
Faf2
Entrez Gene ID
291000
Gene Name
Fas associated factor family member 2
Gene Aliases
2210404d11rik, RGD1306577, Ubxd8
Chromosome Location
Chr.17: 10485650 - 10527882 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006253617.2 XP_006253679.1 2 - 3 185 68
XM_008771488.1 XP_008769710.1 2 - 3 185 68
GenBank mRNA BC079356.1 - 2 - 3 159 68

View Full Product Details