Assay Design



Gene Transcripts

Gene Symbol
Testin
Entrez Gene ID
286916
Gene Name
testin gene
Gene Aliases
-
Chromosome Location
Chr.17: 4194795 - 4225376 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173132.2 NP_775155.1 2 - 3 219 124
XM_008771428.1 XP_008769650.1 3 - 4 579 124
GenBank mRNA U16858.1 - 2 - 3 223 124

View Full Product Details