Assay Design



Gene Transcripts

Gene Symbol
Thoc3
Entrez Gene ID
290519
Gene Name
THO complex 3
Gene Aliases
-
Chromosome Location
Chr.17: 10676943 - 10686134 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106059.1 NP_001099529.1 5 - 6 928 91
GenBank mRNA BC168852.1 - 5 - 6 911 91

View Full Product Details