Assay Design



Gene Transcripts

Gene Symbol
Tspan17
Entrez Gene ID
306771
Gene Name
tetraspanin 17
Gene Aliases
Fbxo23, GHB-R
Chromosome Location
Chr.17: 10356980 - 10364605 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013138.2 NP_001013156.2 5 - 6 728 79
XM_006253630.2 XP_006253692.1 5 - 6 761 79
XM_006253631.2 XP_006253693.1 4 - 5 616 79
GenBank mRNA BC097431.1 - 5 - 6 708 79

View Full Product Details