Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109666.1 | NP_001103136.1 | 8 - 9 | 1325 | 121 |
XM_006235929.2 | XP_006235991.1 | 9 - 10 | 1295 | 121 | |
XM_006235930.2 | XP_006235992.1 | 9 - 10 | 1457 | 121 | |
XM_006235931.2 | XP_006235993.1 | 9 - 10 | 1292 | 121 | |
XM_006235932.2 | XP_006235994.1 | 9 - 10 | 1286 | 121 | |
XM_006235933.2 | XP_006235995.1 | 9 - 10 | 1283 | 121 | |
GenBank mRNA | BC129119.1 | - | 8 - 9 | 1192 | 121 |