Assay Design



Gene Transcripts

Gene Symbol
Crygn
Entrez Gene ID
296730
Gene Name
crystallin, gamma N
Gene Aliases
-
Chromosome Location
Chr.4: 6900907 - 6909167 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106573.1 NP_001100043.1 2 - 3 227 65
XM_006235882.2 XP_006235944.1 3 - 4 898 65

View Full Product Details