Assay Design



Gene Transcripts

Gene Symbol
Smarcd3
Entrez Gene ID
296732
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 3
Gene Aliases
-
Chromosome Location
Chr.4: 7076711 - 7108334 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011966.1 NP_001011966.1 8 - 9 882 93
XM_006235883.2 XP_006235945.1 7 - 8 979 93
XM_006235884.2 XP_006235946.1 7 - 8 979 93
XM_006235885.2 XP_006235947.1 7 - 8 943 93
XM_006235886.2 XP_006235948.1 8 - 9 966 93
XM_006235887.2 XP_006235949.1 8 - 9 930 93
XM_006235888.2 XP_006235950.1 7 - 8 979 93
XM_006235889.2 XP_006235951.1 7 - 8 943 93
GenBank mRNA BC085349.1 - 8 - 9 882 93

View Full Product Details