Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106633.1 | NP_001100103.1 | 2 - 3 | 295 | 138 |
XM_006237805.2 | XP_006237867.1 | 2 - 3 | 320 | 138 | |
GenBank mRNA | FQ214529.1 | - | 2 - 3 | 282 | 138 |
FQ216110.1 | - | 2 - 3 | 311 | 138 | |
FQ216137.1 | - | 2 - 3 | 261 | 138 |