Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106633.1 | NP_001100103.1 | 3 - 4 | 462 | 95 |
XM_006237805.2 | XP_006237867.1 | 3 - 4 | 487 | 95 | |
GenBank mRNA | FQ214529.1 | - | 3 - 4 | 449 | 95 |
FQ216110.1 | - | 3 - 4 | 478 | 95 | |
FQ216137.1 | - | 3 - 4 | 428 | 95 | |
FQ221067.1 | - | 3 - 4 | 447 | 95 |