Assay Design



Gene Transcripts

Gene Symbol
Tox
Entrez Gene ID
362481
Gene Name
thymocyte selection-associated high mobility group box
Gene Aliases
-
Chromosome Location
Chr.5: 19892698 - 20189721 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108654.1 NP_001102124.1 7 - 8 1220 69
XM_006237844.2 XP_006237906.1 7 - 8 1579 69
XM_008763547.1 XP_008761769.1 6 - 7 1325 69

View Full Product Details