Assay Design



Gene Transcripts

Gene Symbol
Pold2
Entrez Gene ID
289758
Gene Name
polymerase (DNA directed), delta 2, accessory subunit
Gene Aliases
-
Chromosome Location
Chr.14: 86111416 - 86117578 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013050.1 NP_001013068.1 2 - 3 298 83
GenBank mRNA BC079267.1 - 2 - 3 298 83

View Full Product Details