Assay Design



Gene Transcripts

Gene Symbol
Klhl8
Entrez Gene ID
289457
Gene Name
kelch-like family member 8
Gene Aliases
-
Chromosome Location
Chr.14: 7169515 - 7215025 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105995.1 NP_001099465.1 8 - 9 1826 85
XM_008770004.1 XP_008768226.1 10 - 11 2161 85
XM_008770005.1 XP_008768227.1 10 - 11 2065 85
XM_008770006.1 XP_008768228.1 10 - 11 2003 85
XM_008770007.1 XP_008768229.1 9 - 10 1830 85

View Full Product Details