Assay Design



Gene Transcripts

Gene Symbol
Parp14
Entrez Gene ID
303903
Gene Name
poly (ADP-ribose) polymerase family, member 14
Gene Aliases
RGD1310490
Chromosome Location
Chr.11: 68105442 - 68137509 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191659.1 NP_001178588.1 8 - 9 3503 92
GenBank mRNA FQ218194.1 - 1 - 2 122 92

View Full Product Details