Assay Design



Gene Transcripts

Gene Symbol
Celf6
Entrez Gene ID
300758
Gene Name
CUGBP, Elav-like family member 6
Gene Aliases
Brunol6
Chromosome Location
Chr.8: 64364497 - 64428452 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106827.1 NP_001100297.1 11 - 12 1401 74
XM_008766227.1 XP_008764449.1 12 - 13 1711 74
XM_008766228.1 XP_008764450.1 12 - 13 1708 74
XM_008766229.1 XP_008764451.1 11 - 12 1648 74
XM_008766230.1 XP_008764452.1 10 - 11 1567 74

View Full Product Details