Assay Design



Gene Transcripts

Gene Symbol
Ccdc12
Entrez Gene ID
363151
Gene Name
coiled-coil domain containing 12
Gene Aliases
RGD1306081
Chromosome Location
Chr.8: 118925681 - 118977038 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108783.1 NP_001102253.1 1 - 2 154 75
XM_006243970.2 XP_006244032.1 2 - 3 778 75

View Full Product Details