Assay Design



Gene Transcripts

Gene Symbol
Elp6
Entrez Gene ID
363150
Gene Name
elongator acetyltransferase complex subunit 6
Gene Aliases
RGD1305478, Tmem103
Chromosome Location
Chr.8: 118525770 - 118541074 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108782.2 NP_001102252.1 5 - 6 546 99
GenBank mRNA BC166773.1 - 5 - 6 546 99

View Full Product Details