Assay Design



Gene Transcripts

Gene Symbol
Cdcp1
Entrez Gene ID
301082
Gene Name
CUB domain containing protein 1
Gene Aliases
RGD1305578
Chromosome Location
Chr.8: 132260007 - 132296661 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106869.1 NP_001100339.1 3 - 4 775 70
XM_006244189.2 XP_006244251.1 3 - 4 766 70

View Full Product Details