Assay Design



Gene Transcripts

Gene Symbol
Mrpl3
Entrez Gene ID
300974
Gene Name
mitochondrial ribosomal protein L3
Gene Aliases
-
Chromosome Location
Chr.8: 113603533 - 113626893 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106852.1 NP_001100322.1 5 - 6 756 94
GenBank mRNA FQ233139.1 - 1 - 2 110 94

View Full Product Details