Assay Design



Gene Transcripts

Gene Symbol
Gsta4
Entrez Gene ID
300850
Gene Name
glutathione S-transferase alpha 4
Gene Aliases
-
Chromosome Location
Chr.8: 85497557 - 85514732 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106840.1 NP_001100310.1 - 610 90
GenBank mRNA BC157819.1 - - 580 90
FJ179399.1 - 5 - 6 543 90
FQ230022.1 - - 590 90
FQ233696.1 - - 592 90
FQ233840.1 - - 592 90
X62660.1 - - 571 90

View Full Product Details