Assay Design



Gene Transcripts

Gene Symbol
Gk5
Entrez Gene ID
367146
Gene Name
glycerol kinase 5 (putative)
Gene Aliases
RGD1311223
Chromosome Location
Chr.8: 103929114 - 104001898 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008766574.1 XP_008764796.1 9 - 10 885 71

View Full Product Details