Assay Design



Gene Transcripts

Gene Symbol
Tbc1d2b
Entrez Gene ID
315880
Gene Name
TBC1 domain family, member 2B
Gene Aliases
RGD1307436
Chromosome Location
Chr.8: 97577551 - 97647072 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108175.1 NP_001101645.1 10 - 11 2111 81
XM_006243528.2 XP_006243590.1 9 - 10 2290 81
XM_008766449.1 XP_008764671.1 9 - 10 2313 81

View Full Product Details