Assay Design



Gene Transcripts

Gene Symbol
Baat
Entrez Gene ID
29725
Gene Name
bile acid CoA:amino acid N-acyltransferase
Gene Aliases
kan-1
Chromosome Location
Chr.5: 64768397 - 64777368 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017300.2 NP_058996.2 1 - 2 145 118
GenBank mRNA BC088153.1 - 1 - 2 145 118
D43964.1 - 1 - 2 118
FQ218773.1 - - 189 118

View Full Product Details