Assay Design



Gene Transcripts

Gene Symbol
Il9
Entrez Gene ID
116558
Gene Name
interleukin 9
Gene Aliases
-
Chromosome Location
Chr.17: 8558827 - 8561950 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105747.1 NP_001099217.1 1 - 2 138 68
GenBank mRNA L36460.1 - 1 - 2 152 68

View Full Product Details