Assay Design



Gene Transcripts

Gene Symbol
Ly96
Entrez Gene ID
448830
Gene Name
lymphocyte antigen 96
Gene Aliases
MD-2
Chromosome Location
Chr.5: 1972212 - 1989448 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024279.1 NP_001019450.1 - 203 103
GenBank mRNA AY963291.1 - - 203 103

View Full Product Details