Assay Design



Gene Transcripts

Gene Symbol
Tceb1
Entrez Gene ID
64525
Gene Name
transcription elongation factor B (SIII), polypeptide 1
Gene Aliases
-
Chromosome Location
Chr.5: 2042991 - 2059357 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006237724.2 XP_006237786.1 1 - 2 76 130

View Full Product Details