Assay Design



Gene Transcripts

Gene Symbol
Ubxn2b
Entrez Gene ID
312965
Gene Name
UBX domain protein 2B
Gene Aliases
RGD1308557
Chromosome Location
Chr.5: 19285224 - 19314199 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107905.1 NP_001101375.1 1 - 2 110 152
XM_006237837.2 XP_006237899.1 1 - 2 128 152
XM_008763541.1 XP_008761763.1 1 - 2 128 152

View Full Product Details