Assay Design



Gene Transcripts

Gene Symbol
Tram1
Entrez Gene ID
312903
Gene Name
translocation associated membrane protein 1
Gene Aliases
-
Chromosome Location
Chr.5: 5055892 - 5064313 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007701.1 NP_001007702.1 - 727 88
GenBank mRNA AY387055.1 - - 610 88
BC083851.1 - - 727 88

View Full Product Details